Digital Microscope Imager, 5 MP
Supplier: Celestron International
44422
470327-778EA 220.25 CAD
470327-778
Digital Microscope Imager, 5 MP
Microscopes
Turns any existing traditional microscope with 23 or 30 mm eyepiece diameters into a digital microscope.
  • Simple, rugged aluminum housed CMOS sensors
  • Takes high resolution still images and 30 fps video
  • USB powered – connects to your personal computer via open USB port
  • Software included
In the classroom, lab, and for any person who has a traditional microscope, Celestron's 5 MP digital microscope imager is the perfect tool to capture high resolution images and 30 fps video. Simply assemble the adjustment collar and the imager will replace any standard 22More Product Information