Blue Phantom® Ultrasound Training Block Models
Supplier: ELEVATE HEALTHCARE, INC. SE
BPAB210 BPO100 BPBV110 BPFB140 BPP120 BPNB150 BPTM130 BPT195
470327-308EA 2225.39 CAD
470327-308 470327-314 470327-316 470327-318 470327-320 470327-372 470327-410 470327-416 470327-426 470327-440
Blue Phantom® Ultrasound Training Block Models
Models Human Biology Models
Compact and convenient.
  • Extremely realistic imaging characteristics
  • Constructed of ultra-durable simulated human tissue, for a realistic training experience
  • Feels and cannulates like real human tissue
  • Self-healing tissue
  • Fluid can be injected into the model to verify needle tip location (automatically expelled)
Blue Phantom Select series vascular access ultrasound training models are an excellent training phantom to help clinicians develop, practice, and maintain the skills necessary for ultrasound guided venous access and arterial access procedures.More Product Information