Celestron StarSense Explorer LT 114AZ Telescope
Supplier: Celestron International
22452
470324-858EA 366.69 CAD
470324-858
Celestron StarSense Explorer LT 114AZ Telescope
Telescopes
StarSense Explorer is ideal for beginners thanks to the app’s user-friendly interface and detailed tutorials.
  • Tour the night sky with your smartphone and the Celestron StarSense Explorer LT 114AZ smartphone telescope
  • Lock targets for eyepiece viewing with in-app bullseye and slow-motion manual controls
  • 114 mm (4.5 inch) Newtonian reflector with highly reflective coatings for excellent viewing
  • Includes 10 mm and 25 mm eyepieces, 2x Barlow lens, smartphone dock, red dot finderscope, and full-height tripod
Leave complicated star charts, impreciseMore Product Information