SIM Prostate Ultrasound And Biopsy Phantom
Supplier: SIMULATED INANIMATE MODELS LLC SE
860000027151 860000027113
470323-456EA 953.05 CAD
470323-456 470323-458
SIM Prostate Ultrasound And Biopsy Phantom
Educational Materials Biology Educational Materials Human Biology Learning Activities
A multi-modality disposable phantom for imaging and biopsy of the prostate.
  • Made of water based hydrogel
  • Imaging compatible
  • Available with or without legions
  • Includes a dicom file
SIM Prostate Phantoms are composed of a water-based hydrogel material that enables superior ultrasound imaging properties versus the competition and are compatible with both standard side-fire and end-fire ultrasound probes. The product is ideal for transrectal ultrasound calibration and training and includes two internal malignant lesions that simulate targeted biopsy andMore Product Information