Formaldehyde-Free Rats
Supplier: Ward's Science
140.1178.1 140.1180.1 140.0948.10 140.0950.10 140.0952.10 470323-096 470323-098 470323-100 470323-102 470323-112
470001-698EA 42.05 CAD
470001-698 470001-708 470000-728 470000-732 470000-736 470323-096 470323-098 470323-100 470323-102 470323-112
Formaldehyde-Free Rats
Specimens Preserved Specimens Preserved Animal Materials
Formaldehyde-free rats. Available in vacuum packs.
  • Mammalian anatomy
  • High quality preservation
To better serve the needs of all teaching environments and preferences, WARD’S offers this line as an alternative to our existing line of high-quality, safe preserved specimens. Absolutely no formaldehyde whatsoever is used in the fixing and preserving process for these specimens. Instead, our new solution used for preparation binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free. WARD’S offersMore Product Information