Media/Storage Bottles, GL 45, Square, Clear Plastic (PC)
Supplier: United Scientific Supplies
BMPC150 BMPC250 BMPC500 BMPC1000
470320-960PK 204.65 CAD
470320-960 470320-962 470320-964 470320-966
Media/Storage Bottles, GL 45, Square, Clear Plastic (PC)
Bottles Plastic Bottles
Clear polycarbonate bottles for storage.
  • Square space-saving design
  • Supplied with polypropylene cap
  • Sterilized using gamma radiation
  • Individually bagged
  • Autoclavable
Clear, square disposable polycarbonate bottles are space-saving and convenient to handle. Useful for mixing, sampling, and storage applications. Graduations and large marking spot are screened in easy-to-read white enamel. Includes a blue, linerless GL 45 polypropylene plug-seal autoclavable cap. Neck int.Ø is 37 mm.
Polycarbonate bottles can be used in environments ranging from −80...+More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable