Thermometer, Digital, Min/Max
Supplier: United Scientific Supplies
DMMT01
470320-734EA 79.15 CAD
470320-734
Thermometer, Digital, Min/Max
Thermometers Digital Thermometers
Easy read digital thermometer.
  • Records the minimum and maximum temperatures for seven days at a time
  • Water-resistant housing + cover for protecting the LCD display
  • Low battery indicator
  • Selectable °C or °F
  • Dual channels with separate temperature readings for each probe
  • Reset button and auto-off display
Sensors: Two with 5’L sensor leads.
Ordering information: Operates on two "AA" batteries (not included).
Order Now
SPECIFICATIONS
Environmentally Preferable