CanDo® Treatment Tables With Adjustable Back
Supplier: Fabrication Enterprises
15-4232(RB-N) 15-4232(GR-N) 15-4232(DT-N) 15-4232(BK-C) 15-4232(SL-C) 15-4232(RB-C) 15-4232(GR-C) 15-4232(DT-C) 15-4233(BK-N) 15-4233(SL-N) 15-4233(RB-N) 15-4233(GR-N) 15-4233(DT-N) 15-4233(BK-C) 15-4233(SL-C) 15-4233(RB-C) 15-4233(GR-C) 15-4233(DT-C) 15-4245(BK-N) 15-4245(SL-N) 15-4245(RB-N) 15-4245(GR-N) 15-4245(DT-N) 15-4245(BK-C) 15-4245(SL-C) 15-4245(RB-C) 15-4245(GR-C) 15-4245(DT-C) 15-4246(BK-N) 15-4246(DT-N) 15-4231(BK-N) 15-4231(SL-N) 15-4231(RB-N) 15-4231(GR-N) 15-4231(DT-N) 15-4231(BK-C) 15-4231(SL-C) 15-4231(RB-C) 15-4231(GR-C) 15-4231(DT-C) 15-4232(BK-N) 15-4232(SL-N) 15-4246(BK-C) 15-4246(SL-C) 15-4246(DT-C) 15-4246(GR-C) 15-4246(GR-N) 15-4246(RB-C) 15-4246(RB-N) 15-4246(SL-N)
470315-872EA 1621.55 CAD
470315-872 470315-874 470315-876 470315-878 470315-880 470315-882 470315-884 470315-886 470315-888 470315-890 470315-892 470315-894 470315-896 470315-898 470315-900 470315-902 470315-904 470315-906 470315-908 470315-910 470315-912 470315-914 470315-916 470315-918 470315-920 470315-922 470315-924 470315-926 470315-928 470315-930 470315-974 470315-976 470315-978 470315-980 470315-982 470315-984 470315-986 470315-988 470315-990 470315-992 470315-994 470315-996 470315-998 470316-000 470316-670 470316-672 470316-674 470316-676 470316-678 470316-680
CanDo® Treatment Tables With Adjustable Back
Furniture Tables and Workbenches
The CanDo® treatment table is a fast and easy to assemble treatment table with a scratch resistant topcoat finish.
  • Five surface and two frame color options
  • Available with or without storage shelf
  • Hardwood legs, a laminate shelf and adjustable backrest for better positioning
  • Topcoat finish is made of a heavyweight, knit-backed upholstery with 2" foam padding and seamless rounded corner top
  • Fixed height
  • Easily assembled
Max load: 400 lbs.
Order Now
SPECIFICATIONS
Environmentally Preferable