ADC® Adscope® 641 Sprague Stethoscopes
Supplier: AMERICAN DIAGNOSTIC CORP CP
641BK
470315-686CS 892.3 CAD
470315-686
ADC® Adscope® 641 Sprague Stethoscopes
Stethoscopes
Why own just a sprague stethoscope when you can own an adscope sprague?
  • Proprietary valve mechanism prevents acoustic leakage
  • Not made with natural rubber latex
The legendary design features of the Sprague have made it one of the most popular stethoscopes in history. ADC®'s Adscope Sprague 641 offers a double-tube design for improved stereo separation, with a 22" tubing length (and a 30" length overall). The threaded chest piece drum has five interchangeable fittings for complete diagnostic capability on most patients.
Delivery information: More Product Information