ADC® Satin Laryngoscopes
Supplier: AMERICAN DIAGNOSTIC CORP CP
4066 4065F 4066F 4067 4070 4072F 4073F 4074F 4080 4081 4082 4082F 4083 4083F 4084 4084F 4501-1 4073D-20 4074D-20 4065D-20 4071D-20 4072D-20
470119-682CS 3863.75 CAD
470119-682 470315-620 470315-622 470315-624 470315-626 470315-628 470315-630 470315-632 470315-634 470315-636 470315-638 470315-640 470315-642 470315-644 470315-646 470315-648 470315-654 470315-764 470315-766 470315-768 470315-772 470315-774
ADC® Satin Laryngoscopes
Educational Materials Biology Educational Materials Human Biology Learning Activities
Compatible with all major brands.
  • Standard, fiber-optic and disposable options
  • Includes one year warranty
Available in standard, fiber-optic, or disposable fiber-optic versions, each is compatible with brands complying with ASTM F965 or ISO 7367. Standard and fiber-optic handles are chrome plated brass, and lightly knurled for a secure grip even while wearing wet gloves. The disposable fiber-optic handle is made of ABS with a ribbed grip. Handles and blades are easily sorted, with fiber-optic version all marked with a green band or insert. ReusableMore Product Information