Ohaus® Starter Pen Meters
Supplier: OHAUS CORP.
30393199 30393200
470315-552EA 725.6 CAD
470315-552 470315-554
Ohaus® Starter Pen Meters
Electrochemistry Meters pH and Multi-Parameter Meters
Compact design for fast, convenient electrochemistry measurement at your fingertips.
  • LCD display
  • Four 1.5 V batteries (included)
  • Easily replaceable electrodes
  • Automatic shut-off
  • Features a 3-point calibration
OHAUS® precision is now available in a line of pen meters that provide accurate measurement of pH, conductivity, salinity, and total dissolved solids (TDS). Starter pen meters are the economical option when you are simply looking for meters that will provide accurate measurement without restriction.
Model ST20M-B: Conductivity Range: 0 – 1999 μs/More Product Information