Adson-Brown Tissue Forceps
Supplier: DR INSTRUMENTS INC. SE
13AB
470315-180BX 390 CAD
470315-180
Adson-Brown Tissue Forceps
Forceps
Easily manipulate delicate tissue.
  • Broad control surface
  • High quality stainless steel
Using narrow tips, and a broad thumb control surface, Adson-Brown forceps give you excellent control when handling delicate tissue. The tips are designed to minimize damage to tissue applying pressure evenly over the full contact surface.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.