Double Burette Clamp with Rod and Stand, Eisco Scientific
Supplier: Wards
CH2019004
470315-090EA 278.1 CAD
470315-090
Double Burette Clamp with Rod and Stand, Eisco Scientific
Clamps General Purpose Clamps
Secure up to two burettes with this complete double burette support clamp and stand set.
  • Double burrete clamp with rod and stand
  • Alloy jaws are suitable for all burettes up to 100mL
  • Cast iron blue base measuring 7" wide and 13" long
  • 12 mm diameter stainless steel support rod measuring 24" long
  • Great for any science classroom or laboratory
Secure one or two burettes with this complete set that includes (1) double burette clamp with boss head, 1 stainless steel rod, and 1 cast iron, epoxy powder coated base. The metal alloy clamp will hold burettes withMore Product Information