Somso® Tulip Bulb
Supplier: MARCUS SOMMER
BOS15/3
470312-444EA 630.4 CAD
470312-444
Somso® Tulip Bulb
Models Botany Models
Somso® premium quality models.
  • 5 year warranty
Tulipa gesneriana, enlarged approx. 5 times, in SOMSO-Plast®. After Prof. Dr. W. Weber. The model shows the longitudinal section of a shooting tulip bulb. Separable into 3 parts. On a green base with key.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.