Somso® Lower Plants
Supplier: MARCUS SOMMER
BOS14/2
470312-424EA 541.3 CAD
470312-424
Somso® Lower Plants
Models Botany Models
Somso® premium quality models.
  • 5 year warranty
Marchantia polymorpha. Antheridium, enlarged approx. 1500 times, in SOMSO-Plast®. After Prof. Dr. W. Weber. In one piece. On a green base with explanatory note.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.