Ward's Benchtop Counter
Supplier: VWR International
HS5360AW HS5360BW
470311-184CS 6873 CAD
470311-184 470311-186
Ward's Benchtop Counter
Cell Counters and Flow Cytometers Manual Cell Counters
Manual counting made simple.
  • Each key records up to 999
  • Six and nine key options available
Ward's mechanical, benchtop counters are perfect for blood cells (label included), bacterial colonies, or other repetitive counting. Total window keeps track of the total number of strokes made on all other keys. A bell rings when total reaches 100 Blood cell key labels. 6-Key: baso., eosin., mono., lymph., poly., and total; 9-Key: baso., eosin., myelo., juven., stab., seg., lymph., mono., and total.
Order Now
SPECIFICATIONS
Environmentally Preferable