Klinger Montmorillonite Crystal Model
Supplier: Klinger
KS7083
470311-146EA 3309.35 CAD
470311-146
Klinger Montmorillonite Crystal Model
Models Crystal Models
This model is hand made in the USA by Klinger Educational Products.
  • Contains 252 - 1" balls
This is a permanent structure. We only use grade A materials. The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.