Ward's® Build Your Own Insect Net
Supplier: Shiv Dial Sud & Sons
B11235H/3 B11235R/1 B11235R/2 B11235R/3 B11235R/4 B11235R/5 B11235R/6 B11235R/7 B11235N/1 B11235N/2 B11235N/3 B11235N/4 B11235N/5 B11235N/6 B11235N/7 B11235N/8 B11235H/1 B11235N/4-100
470308-192CS 1442.9 CAD
470308-192 470308-196 470308-198 470308-200 470308-202 470308-204 470308-206 470308-208 470308-210 470308-212 470308-214 470308-216 470308-218 470308-220 470308-222 470308-224 470308-262 470347-472
Ward's® Build Your Own Insect Net
Animal Supplies Animal Nets
Customize your own insect net, or replace existing parts.
  • Hardwood or aluminum handles
  • Zinc-plated steel rings are corrosion-resistant
  • Aquatic and non-aquatic options
  • Options for younger students to adults
If you are looking for a specific net combination, or just looking to expand your collection net options, these components are just what you need. For over 60 years the Ward's component net system has been a favorite of teachers and collectors alike. Now you can build a net in your own custom configuration. Each net is made of three components: aMore Product Information