Medline® Wall Mounted Diagnostic Set
Supplier: MEDLINE INDUSTRIES, INC.
MDS36533002
470303-254EA 2783.95 CAD
470303-254
Medline® Wall Mounted Diagnostic Set
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
A necessity for exam training.
  • LED illumination for otoscope, and xenon for ophthalmoscope
  • Wall mounted convenience
  • Long three-meter spiral instrument cord
The wall mounted diagnostic set can be conveniently placed in exam rooms and expanded to the left or right as desired. This kit includes a power supply, a LED Otoscope and a Xenon Ophthalmoscope. Instruments automatically switch on when removed from the cradle, and off when replaced. A consistent power source extends bulb life and removes the need for constant battery changes.