Methyl Violet 2B

Supplier: ALDON CORP SE

MM0550-100ML
470301-798EA 19.1 CAD
470301-798
Methyl Violet 2B
Chemicals for Science Education

CAS Number: 8004-87-3
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): Approximately 1
Boiling Point (°C): Approximately 100
Freezing Point (°C): Approximately 0
Solubility: Water and Chloroform
Synonyms: Basic Violet 1 Aqueous Solution
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR