Manganese(II) Sulfate Monohydrate

Supplier: ALDON CORP SE

MM0230-100G MM0230-500G MM0230-100GR MM0230-500GR
470301-712EA 22.85 CAD
470301-712 470301-714 470301-716 470301-718
Manganese(II) Sulfate Monohydrate
Chemicals for Science Education

CAS Number: 10034-96-5
Formula Weight: 169.01
Formula: MnSO4·H2O
Hazard Info: Irritant
Density (g/mL): 2.95
Freezing Point (°C): 57-117
Solubility: Water
Synonyms: Manganous Sulfate, Manganese Sulfate
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR