D(+)Maltose Monohydrate

Supplier: ALDON CORP SE

MM0180-500G MM0180-100G
470301-443EA 77.9 CAD
470301-443 470301-704
D(+)Maltose Monohydrate
Chemicals for Science Education

CAS Number: 6363-53-7
Formula Weight: 360.32
Formula: C12H22O11·H2O
Boiling Point (°C): Decomposes
Freezing Point (°C): 102-103
Solubility: Water and Alcohol
Synonyms: Maltose, Malt Sugar
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR