Microscope Lens Cleaning Solution

Supplier: Ward's Science
LL0158-500ML
470301-560EA 91.8 CAD
470301-560
Microscope Lens Cleaning Solution
Cleaning Agents Lens Cleaning Solutions
Density (g/mL): 0.97
Synonyms: Lens Cleaner, Lens Cleaning Solution, Microscope Lens Solution
Shelf Life (months): 36
Storage: Red

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR