Cobalt Sulfate Heptahydrate

Supplier: ALDON CORP SE

CC0365-100G
470300-778EA 139.35 CAD
470300-778
Cobalt Sulfate Heptahydrate
Chemicals for Science Education

CAS Number: 10026-24-1
Formula Weight: 281.10
Formula: CoSO4·7H2O
Hazard Info: Irritant
Density (g/mL): 1.95
Freezing Point (°C): 96.8
Synonyms: Cobaltous Sulfate, Cobalt Sulfate
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR