Carson HookUpz Universal Smartphone Binocular Adapter
Supplier: CARSON OPTICAL
IB-700
470235-860EA 67.95 CAD
470235-860
Carson HookUpz Universal Smartphone Binocular Adapter
Monoculars and Binoculars
The HookUpz Universal Smartphone Binocular Adapter from Carson easily connects your smartphone to most full sized binoculars.
  • Objective Lens Diameter: 32 to 50 mm
Use your built-in camera application on your smartphone to take pictures and record video through the lenses of your binoculars.
Fits all phones. For specific information on your model, visit: www.carson.com/multicam.
Order Now
SPECIFICATIONS
Environmentally Preferable