Ultra Flex Support System
Supplier: OHAUS CORP.
30400011 30400014 30400013 30400016 30400012 30400015
470235-400EA 678.7 CAD
470235-400 470235-402 470235-404 470235-406 470235-408 470235-410
Ultra Flex Support System
Supports Flask Supports
Provides Secure Support for Holding Glassware at Elevated Positions.
  • Choose the configuration that best suits your application
  • Flexible and easy to apply to your specific needs
Ultra Flex Support Systems have unique flexible arms that are extremely versatile and can be placed in virtually any position or angle. Ultra Flex comes in three different systems; base plate, lab-frame connector; and bench clamp. Each system includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring Clamp Head, nickel-plated flex arm in 305 mm (12") or 457 mm (18") length, andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable