OHAUS® Heavy Duty Vortex Mixer
Supplier: OHAUS CORP.
30392146 30400216 30400226 30400217 30400220 30400221 30400222 30400223 30400224 30400210 30400218 30400211 30400215 30400213 30400214 30400212 30400209
470235-166EA 2325.55 CAD
470235-166 470235-362 470235-364 470235-366 470235-368 470235-370 470235-372 470235-374 470235-376 470235-380 470235-382 470235-384 470235-386 470235-388 470235-390 470235-392 470235-394
OHAUS® Heavy Duty Vortex Mixer
Shakers Vortex Mixers
Designed for Continuous Duty Applications.
  • LED displays for speed and time
The OHAUS Digital Heavy-Duty Vortex Mixer is ideal for applications that demand repeatable results. Mixer features touchpad controls and LED displays for accurate speed (rpm) and time (minutes and seconds) results. The heavy-duty design and efficient motor allow this mixer to operate in continuous duty and handle all accessories over the entire speed range. Choose from two modes of operation- "Touch" mode for mixing tubes when cup head or Universal Holder with cover isMore Product Information