OHAUS® Light Duty Orbital Shakers
Supplier: OHAUS CORP.
30391924 30391938 30391931 30391945 30400157 30400159 30400191 30400172 30400168 30400158 30400151 30400152 30400156 30400193 30400194 30400182 30400183 30400190 30400184 30400185 30400186 30400187 30400188 30400189 30400114 30400084 30400085 30400086 30400087 30400088 30400099 30400100 30400115 30400116 30400119 30400120
470235-156EA 5775.75 CAD
470235-156 470235-158 470235-160 470235-162 470235-198 470235-200 470235-202 470235-204 470235-206 470235-208 470235-210 470235-212 470235-214 470235-216 470235-218 470235-220 470235-222 470235-224 470235-226 470235-228 470235-230 470235-232 470235-234 470235-236 470235-288 470235-292 470235-294 470235-296 470235-298 470235-300 470235-322 470235-324 470235-332 470235-334 470235-340 470235-342
OHAUS® Light Duty Orbital Shakers
Shakers Shaking Incubators
Save Incubator Space with Benchtop Orbital Shakers.
  • LED displays for temperature, speed and time
  • Timer with audible alarm
  • Calibration mode for temperature
  • Microprocessor-controlled for consistent shaking action
  • Triple eccentric drive provides reliable service and continuous duty
  • Safety features include speed ramping and overload protection
Ohaus Incubating Light Duty Shakers are designed to incubate samples from 10° below ambient to 65 °C depending on the model. The incubating Mini shaker has a 3.6 kg (8 lb.) capacity while the Incubating MicroplateMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable