Ward's® myVolt™ Mini Power Supply
Supplier: VWR International
470231-452
470231-452EA 440.8 CAD
470231-452
Ward's® myVolt™ Mini Power Supply
Power Supplies
Ideal for basic, constant voltage applications.
  • Compact and simple
  • Chose constant 100 V or 200 V output
  • Two pairs of output terminals
  • Stackable design
  • 2 year warranty
The myVolt Mini is a compact electrophoresis power supply ideal for basic, constant voltage applications. Users can select constant 100 V or 200 V output, and the current can reach a maximum of 200 mA. Two sets of output jacks are available to connect up to two gel rigs.