Ward's® Essentials Spacesuit Design Challenge
Supplier: Science First
650-1000
470231-276EA 80.99 CAD
470231-276
Ward's® Essentials Spacesuit Design Challenge
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
The Perfect Introduction to Temperature and Conduction.
  • Investigation includes teaching guides
  • Students apply standard engineering design processes while applying economics
  • Aligned with NGSS MS-PS3-3, MS-ETS1-1, MS-ETS1-2, MS-ETS1-3, MS-ETS1-4
Students are presented with a defined set of materials and asked to design a spacesuit that can help maintain the body temperature of an astronaut in the extreme heat and cold of outer space. Water-filled “Space Worms” are used as test subjects that mimic the water composition of the adult human body. StudentsMore Product Information