Self-Laminating GHS Tags,
Supplier: EMEDCO
GHSLT
470231-142EA 15.75 CAD
470231-142
Self-Laminating GHS Tags,
Labels
Safeguard important chemical hazard information.
  • Plastic tag can withstand daily wear and tear
  • Attach mini-pictogram labels (sold separately) to recognize hazards
Protect your GHS tags from dirt and grime by peeling off the adhesive layer and sticking it to the front of the tag. Add any additional information to the back of the safety tags.
Certifications: Complies with OSHA 29 CFR 1910.1200 regulation