Color Filter, Plastic

Supplier: Wards
PH0615AD
470230-210EA 10.45 CAD
470230-210
Color Filter, Plastic
Educational Materials Physics Educational Materials Optics Learning Activities
Spare color filter set to be used with Light Box and Optical Set.

  • Spare set of light filters for PH0615 Light Box and Optical Set

Set of 8 plastic color filters for use with Labs Light Box and Optical Set (PH0615). Great as a back up set for when one goes missing. Red, Cyan, Violet, Green, Blue, Yellow, Orange, Magenta
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR