Neodymium Disk Magnet
Supplier: Master Magnetics
ND030N-35 ND143N-35BXX3
470229-346EA 55.2 CAD
470229-346 470229-348 470229-350
Neodymium Disk Magnet
Magnets
A powerful magnet for a variety of experiments.
  • Very high resistance to demagnetization
  • High energy for size
  • Good in ambient temperature
Neodymium Disk Magnets (Nd-Fe-B) are made from the most powerful magnetic material in the world. They boast extreme strength in a small size. All of the magnets have a grade of 35 with a Gauss rating of 12,200. Maximum operating temperature is 80°C (176°F).