LaMotte® Macrolens
Supplier: LaMotte
5508-10
470229-160PK 314.4 CAD
470229-160
LaMotte® Macrolens
Educational Materials Environmental Educational Materials Environmental Learning Activities
Handy for observising and identifying macroinvertabrates.
MacroLens with 4.5X magnification fits over the entire petri dish. 4" diameter. Available in a convenient pack of 10.
Order Now
SPECIFICATIONS
Environmentally Preferable