Component Holder
Supplier: Shiv Dial Sud & Sons
PH60106
470225-196CS 607.5 CAD
470225-196
Component Holder
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Makes hard to connect components easy to use in electrical circuits.
  • With 4 mm socket connectors
The component holder is an instrument for holding the electronic components such as resistors, capacitors, LEDs, LDRs etc for the use in different experimental circuits.