Flying Pig
Supplier: ARBOR SCIENTIFIC
P4-2165
470223-114EA 29.69 CAD
470223-114
Flying Pig
Educational Materials Physics Educational Materials Mechanics Learning Activities
Make students learn when pigs fly.
  • Includes mounting string
  • Includes instruction manual
Add some excitement to your lessons on circular motion with the Flying Pig. Suspended from the ceiling, his wings flap and carry him in a regular circle. With the string, this motion gives students a conical pendulum and a perfect opportunity for quantitative measurements of circular motion.
Ordering information: Lab instructions included.