Rod and Base Assembly
Supplier: VWR International
470220-780 CH0653BRD2 470220-784 470220-786
470220-780EA 36.8 CAD
470220-780 470220-782 470220-784 470220-786
Rod and Base Assembly
Retort Stands
This assembly is handy for multiple lab applications.
  • Heavy-duty cast iron construction
  • Rod thread length: 10 mm
  • Rod thread width: 1.5 mm
  • Blue epoxy powder coating
The base is constructed of heavy-duty cast iron with blue epoxy powder coating.