Rod Friction Set
Supplier: Wards
PH1120150
470219-536EA 49.45 CAD
470219-536
Rod Friction Set
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Economical set for studying friction.
  • Exploration into electrostatic forces
This electrostatic kit provides the necessary tools to free both positive and negative charges for exploration into electrostatic forces. Students can experiment with different combinations of cloth and rod materials to explore how each manufactures charge carriers.
Instruction manual included.
Kit includes:
3 Fabric Cloths (wool, cotton, silk)
3 Rods (glass, ebonite, Lucite)
Order Now
SPECIFICATIONS
Environmentally Preferable