Rolling Friction Car Kit
Supplier: Wards
PH1120147
470219-534EA 108.85 CAD
470219-534
Rolling Friction Car Kit
Educational Materials Physics Educational Materials Applied Physics Learning Activities
The rolling friction car kit provides four cars for exploration into the force of friction. Each car provides a set of different material wheels (wood, fabric, soft rubber, soft silicone) with a different coefficient of friction. By setting the cars in motion with equivalent velocities, one can qualitatively demonstrate the frictional properties of rolling without slipping.
Instruction manual included.