Demonstration Transformer Kit
Supplier: Wards
PH0849AK1 PH0849GT
470219-516EA 185.5 CAD
470219-516 470219-518
Demonstration Transformer Kit
Transformers
The Demonstration Transformer Kit is a safe and convenient system for AC and DC experiments up to 0.5 amps.
  • For AC and DC Experiments up to 0.5A
  • Large-Scale Transformer
  • Ideal for the Classroom
  • Intermediary Outputs
  • Safe and Convenient
The large-scale transformer is ideal for the classroom.
The coils are fitted with intermediary outputs that provide a variety of transformation ratios.
Accessories information: The accessory kit demonstrates how step down transformers can produce currents that are large enough to weld metals or boil water.