Truss Kit
Supplier: Wards
PH0344SFT
470219-430EA 147.55 CAD
470219-430
Truss Kit
Educational Materials Physics Educational Materials Mechanics Learning Activities
Studying dynamic tension has never been easier with this simple form truss kit. The kit includes a half-meter truss with hooks on both sides, V-shaped clamp.Recommended stand and rod, item 470137-892 Required but not included are masses and spring scale.