Corning® PC-200 Low Profile Hot Plate
Supplier: Corning
6795-200
470211-556EA 710.49 CAD
470211-556
Corning® PC-200 Low Profile Hot Plate
Heaters Hot Plates
Low-profile hot plates with a ceramic glass plate provide precision-controlled heating for laboratory applications. Microprocessor-controlled heating ensures consistent performance.
  • Space saving design
  • Hot top indicator light
  • Large knobs provide control for gloved fingers
Hot top indicator light blinks a warning if the top is too hot to touch (>140°F) even when the unit is turned off. Flatter, whiter Pyroceram® top conducts heat more efficiently and evenly. Built-in holder accommodates optional 4 inch stainless steel support rods.
Certifications:More Product Information