Kinetic Lab Tables
Supplier: Diversified Woodcrafts
C2202K C2204K C2302K C2402K C2304K C2404K C2306K C2406K
470205-244EA 3755.2 CAD
470205-244 470205-252 470205-270 470205-274 470205-278 470205-282 470205-286 470205-290
Kinetic Lab Tables
Furniture Tables and Workbenches
Storage cabinet tables constructed of solid oad and hardwood veneers featuring a center cabinet with lock and rubber base molding.
  • 1" epoxy resin or 1¼" ChemGuard work surface
  • Center cabinet with adjustable shelves and locking doors
  • Rubber base molding and leg boots
  • Chemical resistant UV finish
These tables are designed for use at sitting height. They are finished with a chemical resistant, earth-friendly UV finish.
Certifications: Diversified Woodcrafts is SEFA compliant and MAS Certified Green.
Delivery information: Shipping weight is 370 lbs.
Order Now
SPECIFICATIONS
Environmentally Preferable