Valor™ 3000 Xtreme Compact Scales, Ohaus®
Supplier: OHAUS CORP.
83998130
470203-774EA 1691.5 CAD
470203-774
Valor™ 3000 Xtreme Compact Scales, Ohaus®
Balances and Scales
Valor™ 3000 Xtreme compact scales are rugged with a full stainless steel design in a high-performance portable scale.
  • One-year warranty
This compact, durable scale is ideal for dry applications. The high-quality stainless steel construction allows for easy cleaning and adjustable feet keep the scales sturdy. Features include a bright backlit LCD display, percentage weighing and specific gravity software, and an AC adapter. Overload protection is rated at 10X. NSF certified.
Accessories information: Operates on four C batteries, not included. WeightMore Product Information