Beaker Brush
Supplier: Mill-Rose
72110B
470203-408EA 23.75 CAD
470203-408
Beaker Brush
Brushes
Four rows of black pig hair bristles help you clean beakers, funnels, cylinders and other labware. The bristles are twisted into galvanized wire and mounted on a hardwood handle (16 mm diameter).
Overall length: 33cm.