Justrite® Sure-Grip® EX Flammable Safety Cabinet
Supplier: Justrite
893080 893020 894500
470106-728EA 3142.05 CAD
470106-728 470202-962 470202-966
Justrite® Sure-Grip® EX Flammable Safety Cabinet
Furniture Storage Cabinets
Designed to Meet OSHA and NFPA Standards.
  • Feature Haz-Alert™ reflective warning labels
  • Come with cylinder lock and key and accepts additional padlock
  • Shelves designed to protect against spills
  • Also feature dual vents with flame arresters, adjustable feet, grounding connector, and trilingual warning labels
Cabinets are constructed of sturdy 18-gauge (1-mm) thick double-wall, welded steel with 1-1/2” (38-mm) of insulating air space for fire resistance. Manual-close door(s) opens to a full 180 degrees and self-latches when pushed closed. Self-close door(More Product Information