Slide Tray Cabinet
Supplier: Bel-Art
H44581-0000_SE H44583-0000_SE
470157-670CS 1528.5 CAD
470157-670 470201-796
Slide Tray Cabinet
Microscope Slide Boxes
Store your slides securely with a minimal surface footprint.
  • Store slides horizontally for the best care of whole mounts
  • Clear door allows for internal viewing
Our polypropylene microscope slide tray cabinet holds up to 240 slides total. It can accommodate 12 horizontal slide trays in all. The clear acrylic door slides vertically. Slide trays are available separately.