Tergazyme® Enzyme-Active Powdered Detergent
Supplier: Alconox
1304
470201-490CS 975.75 CAD
470201-490
Tergazyme® Enzyme-Active Powdered Detergent
Cleaning Agents Detergents and Surface Cleansers
Tergazyme is a concentrated, anionic detergent with protease enzyme for manual and ultrasonic cleaning. Excellent for removal of proteinaceous soils, tissue, blood and body fluids from glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass with no interfering residues. Ideal as a cleaning agent in hospital instruments, dairy equipment, laboratory ware, reverse osmosis and ultrafiltration membranes and units, sampling apparatus, pharmaceutical apparatus, cosmetics manufacturing equipment, tubing, pipes, optical parts, process equipment, industrial parts, desalination plants,More Product Information