Glass Microfiber Filter Paper
Supplier: AHLSTROM MUNKSJO FILTRATION LLC.
1610-1100
470201-466PK 210.35 CAD
470201-466
Glass Microfiber Filter Paper
Filters Filter Papers Qualitative Filter Paper
The glass filter paper is excellent for microwave testing where temperatures get very high such as for use in a food experiment where all the moisture is microwaved out to see how much solid is left behind. Withstands temperatures to 500 °C. Size of each: 11 cm diameter.
Packaging: Package of 100.