Spinal Cord with Ganglion Slide
Supplier: Avantor
470189-622EA 11.75 CAD
470189-622
Spinal Cord with Ganglion Slide
Slides Prepared Slides
Illustrates components of nerves and nerve tissue
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Shows dorsal and ventral nerve roots and ganglion. Stained for clear distinction of cellular elements, nerve fibers, and connective tissue.
Order Now
SPECIFICATIONS
Environmentally Preferable